Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
N4BP3
Entrez Gene ID
513032
Gene Name
NEDD4 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 39210669 - 39222836 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205742.1 NP_001192671.1 3 - 4 1109 60

View Full Product Details