Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD9
Entrez Gene ID
514205
Gene Name
sterile alpha motif domain containing 9
Gene Aliases
-
Chromosome Location
Chr.4: 10449812 - 10468917 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205781.3 NP_001192710.2 2 - 3 415 90
GenBank mRNA BC151751.1 - 2 - 3 417 90

View Full Product Details