Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP43
Entrez Gene ID
518801
Gene Name
cilia and flagella associated protein 43
Gene Aliases
WDR96
Chromosome Location
Chr.26: 24695313 - 24795331 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205849.3 NP_001192778.3 26 - 27 3414 87

View Full Product Details