Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNE3
Entrez Gene ID
520207
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, C21H14orf49
Chromosome Location
Chr.21: 60265774 - 60379931 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205869.2 NP_001192798.1 13 - 14 2298 92

View Full Product Details