Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VANGL2
Entrez Gene ID
520544
Gene Name
VANGL planar cell polarity protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 9263557 - 9291287 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205875.1 NP_001192804.1 5 - 6 1385 60

View Full Product Details