Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP70
Entrez Gene ID
529891
Gene Name
cilia and flagella associated protein 70
Gene Aliases
TTC18
Chromosome Location
Chr.28: 29299960 - 29383242 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205969.1 NP_001192898.1 22 - 23 2897 87

View Full Product Details