Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT5A
Entrez Gene ID
530005
Gene Name
Wnt family member 5A
Gene Aliases
-
Chromosome Location
Chr.22: 45540765 - 45561515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205971.1 NP_001192900.1 2 - 3 700 94

View Full Product Details