Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCU
Entrez Gene ID
536656
Gene Name
mitochondrial calcium uniporter
Gene Aliases
CCDC109A
Chromosome Location
Chr.28: 28709011 - 28904117 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206102.1 NP_001193031.1 2 - 3 228 100

View Full Product Details