Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTR3C
Entrez Gene ID
519394
Gene Name
5-hydroxytryptamine (serotonin) receptor 3C, ionotropic
Gene Aliases
-
Chromosome Location
Chr.1: 83064169 - 83070990 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192508.1 NP_001179437.1 5 - 6 560 74

View Full Product Details