Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGCS1
Entrez Gene ID
407767
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 1
Gene Aliases
-
Chromosome Location
Chr.20: 31431132 - 31456871 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206578.1 NP_001193507.1 7 - 8 1296 66
GenBank mRNA BC102850.1 - 7 - 8 1206 66
EE350662.1 - 4 - 5 551 66

View Full Product Details