Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNOT8
Entrez Gene ID
533443
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
-
Chromosome Location
Chr.7: 66042162 - 66061078 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192774.1 NP_001179703.1 5 - 6 823 85
GenBank mRNA BC119978.1 - 6 - 7 1071 85

View Full Product Details