Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC69
Entrez Gene ID
529878
Gene Name
coiled-coil domain containing 69
Gene Aliases
-
Chromosome Location
Chr.7: 62434491 - 62469922 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206624.1 NP_001193553.1 1 - 2 161 74
GenBank mRNA BC148945.1 - 1 - 2 161 74

View Full Product Details