Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF12
Entrez Gene ID
525161
Gene Name
DDB1 and CUL4 associated factor 12
Gene Aliases
WDR40A
Chromosome Location
Chr.8: 75491759 - 75531993 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167896.2 NP_001161368.1 1 - 2 385 101
GenBank mRNA BC105241.1 - 1 - 2 76 101
BC150062.1 - 1 - 2 384 101

View Full Product Details