Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTE3
Entrez Gene ID
789619
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
-
Chromosome Location
Chr.9: 103525415 - 103548203 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191253.1 NP_001178182.1 3 - 4 475 112

View Full Product Details