Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UST
Entrez Gene ID
535975
Gene Name
uronyl 2-sulfotransferase
Gene Aliases
-
Chromosome Location
Chr.9: 86010485 - 86327506 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206826.1 NP_001193755.1 2 - 3 302 84
NM_001206827.1 NP_001193756.1 2 - 3 302 84
GenBank mRNA DV895087.1 - 1 - 2 43 84

View Full Product Details