Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGLON5
Entrez Gene ID
786369
Gene Name
IgLON family member 5
Gene Aliases
-
Chromosome Location
Chr.18: 57390098 - 57404602 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206875.1 NP_001193804.1 3 - 4 394 73
GenBank mRNA EE351204.1 - 1 - 2 46 73
EH200051.1 - 3 - 4 371 73

View Full Product Details