Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM240
Entrez Gene ID
617545
Gene Name
transmembrane protein 240
Gene Aliases
C16H1orf70, C1orf70
Chromosome Location
Chr.16: 51079295 - 51084984 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206963.1 NP_001193892.1 3 - 4 372 109
GenBank mRNA EV628200.1 - 3 - 4 372 109

View Full Product Details