Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H19orf33
Entrez Gene ID
787634
Gene Name
chromosome 18 C19orf33 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 48118686 - 48119892 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207046.1 NP_001193975.1 2 - 3 232 80
GenBank mRNA BC102086.1 - 2 - 3 233 80
EV616576.1 - 2 - 3 183 80
EV688939.1 - 2 - 3 137 80

View Full Product Details