Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC10
Entrez Gene ID
100138038
Gene Name
armadillo repeat containing 10
Gene Aliases
-
Chromosome Location
Chr.4: 44363535 - 44500937 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191332.3 NP_001178261.1 3 - 4 789 83

View Full Product Details