Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIT1
Entrez Gene ID
538858
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
Gene Aliases
-
Chromosome Location
Chr.28: 39201718 - 39213131 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192115.1 NP_001179044.1 3 - 4 899 57

View Full Product Details