Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNRF1
Entrez Gene ID
509076
Gene Name
zinc and ring finger 1
Gene Aliases
-
Chromosome Location
Chr.18: 2362997 - 2451045 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192210.1 NP_001179139.1 1 - 2 547 60

View Full Product Details