Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT84
Entrez Gene ID
539944
Gene Name
keratin 84
Gene Aliases
-
Chromosome Location
Chr.5: 27506039 - 27512835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192335.1 NP_001179264.1 3 - 4 785 64

View Full Product Details