Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNB3
Entrez Gene ID
513340
Gene Name
G protein subunit beta 3
Gene Aliases
-
Chromosome Location
Chr.5: 103602563 - 103609014 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192347.1 NP_001179276.1 9 - 10 1033 69

View Full Product Details