Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL26A1
Entrez Gene ID
617340
Gene Name
collagen type XXVI alpha 1 chain
Gene Aliases
EMID2
Chromosome Location
Chr.25: 35219723 - 35383571 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193154.1 NP_001180083.1 3 - 4 502 56

View Full Product Details