Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COX4I2
Entrez Gene ID
618339
Gene Name
cytochrome c oxidase subunit 4I2
Gene Aliases
-
Chromosome Location
Chr.13: 61200040 - 61205420 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193186.1 NP_001180115.1 4 - 5 451 66

View Full Product Details