Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCR8
Entrez Gene ID
407772
Gene Name
C-C motif chemokine receptor 8
Gene Aliases
-
Chromosome Location
Chr.22: 12590658 - 12636482 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194962.1 NP_001181891.1 1 - 1 1018 66
GenBank mRNA GU936963.1 - 1 - 1 1018 66

View Full Product Details