Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALM2
Entrez Gene ID
100532750
Gene Name
paralemmin 2
Gene Aliases
AKAP2, PALM2-AKAP2, PALM2AKAP2
Chromosome Location
Chr.8: 99342276 - 99503025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204119.1 NP_001191048.1 6 - 7 499 84
GenBank mRNA EH372970.1 - 6 - 7 513 84

View Full Product Details