Product Details

SNP ID
rs80179427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112041576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGAATCAAAATAAATTACCAT[G/T]TATTCCTTCAGAGAGCCCATAAAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS7 PubMed Links

Gene Details

Gene
TMPRSS7
Gene Name
transmembrane protease, serine 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042575.2 Intron NP_001036040.2
XM_011512754.1 Intron XP_011511056.1
XM_011512758.1 Intron XP_011511060.1

View Full Product Details