Product Details

SNP ID
rs79178079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179246790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCGAGTACTGAACTGAGTATGA[C/T]TTGAATTTTCCTTAAAGATTTTAGA
Phenotype
MIM: 605222 MIM: 171834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNMB3 PubMed Links

Gene Details

Gene
KCNMB3
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163677.1 Intron NP_001157149.1
NM_014407.3 Intron NP_055222.3
NM_171828.2 Intron NP_741979.1
NM_171829.2 Intron NP_741980.1
NM_171830.1 Intron NP_741981.1
Gene
PIK3CA
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
There are no transcripts associated with this gene.

View Full Product Details