Product Details

SNP ID
rs74725336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43337060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGCTGACTGCATCTCTTGGTG[C/T]AGGTTCCAGATGAGATAACAAACTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF318 PubMed Links

Gene Details

Gene
ZNF318
Gene Name
zinc finger protein 318
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014345.2 7016 UTR 3 NP_055160.2
XM_011514451.2 7016 Intron XP_011512753.1
XM_011514452.2 7016 Intron XP_011512754.1

View Full Product Details