Product Details

SNP ID
rs76444240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47658384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGATTTTATTTTTATTTAGGGA[C/T]GAGGCTATAGAACCTACTGCTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRF2 PubMed Links

Gene Details

Gene
ADGRF2
Gene Name
adhesion G protein-coupled receptor F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153839.6 Intron NP_722581.4

View Full Product Details