Product Details

SNP ID
rs76790123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77311627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAACATTTGCATCAACCAAATGG[A/G]CTTTAAACCTCTTGACTCTTTTAAA
Phenotype
MIM: 613552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSAP PubMed Links

Gene Details

Gene
GSAP
Gene Name
gamma-secretase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017439.3 Intron NP_059135.2
XM_011516325.2 Intron XP_011514627.1
XM_011516326.2 Intron XP_011514628.1
XM_011516327.2 Intron XP_011514629.1
XM_011516330.2 Intron XP_011514632.1
XM_011516331.2 Intron XP_011514633.1
XM_011516332.2 Intron XP_011514634.1
XM_017012343.1 Intron XP_016867832.1
XM_017012344.1 Intron XP_016867833.1
XM_017012345.1 Intron XP_016867834.1
XM_017012346.1 Intron XP_016867835.1
XM_017012347.1 Intron XP_016867836.1
XM_017012348.1 Intron XP_016867837.1
XM_017012349.1 Intron XP_016867838.1
XM_017012350.1 Intron XP_016867839.1
XM_017012351.1 Intron XP_016867840.1

View Full Product Details