Product Details

SNP ID
rs79344159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:141137447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTTAAAATGAAGTCTTGCAGT[A/G]GTTGGCAGATGGATTAAAAATACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND3 PubMed Links

Gene Details

Gene
DENND3
Gene Name
DENN domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014957.2 Intron NP_055772.2
XM_005250838.4 Intron XP_005250895.2
XM_005250839.2 Intron XP_005250896.2
XM_005250840.4 Intron XP_005250897.2
XM_005250841.3 Intron XP_005250898.2
XM_005250842.4 Intron XP_005250899.1
XM_011516933.1 Intron XP_011515235.1
XM_011516934.2 Intron XP_011515236.1
XM_011516937.1 Intron XP_011515239.1
XM_011516938.2 Intron XP_011515240.1
XM_011516939.2 Intron XP_011515241.1
XM_011516940.2 Intron XP_011515242.1
XM_011516941.2 Intron XP_011515243.1
XM_017013241.1 Intron XP_016868730.1
XM_017013242.1 Intron XP_016868731.1
XM_017013243.1 Intron XP_016868732.1

View Full Product Details