Product Details

SNP ID
rs78450021
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122788947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAGTAGGATGAACTCAGAGAA[C/G]CTCACCCGGGCCGCGGTTGCCCCTG
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR5C1 PubMed Links

Gene Details

Gene
OR5C1
Gene Name
olfactory receptor family 5 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001923.1 15 Missense Mutation AAC,AAG N5K NP_001001923.1

View Full Product Details