Product Details

SNP ID
rs74389187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18231537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGCAGGCCGTCAGGTCTGAAGC[A/G]GTCGGGGTCTTTGCCACTCCGGCCC
Phenotype
MIM: 104751 MIM: 104752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAA2 PubMed Links

Gene Details

Gene
SAA2
Gene Name
serum amyloid A2
There are no transcripts associated with this gene.

Gene
SAA2-SAA4
Gene Name
SAA2-SAA4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199744.1 668 Missense Mutation CGC,TGC R198C NP_001186673.1
Gene
SAA4
Gene Name
serum amyloid A4, constitutive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006512.3 668 Missense Mutation CGC,TGC R120C NP_006503.2

View Full Product Details