Product Details

SNP ID
rs79025622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3361333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCAAAGATGACATGTACAAAG[C/T]ATTAGGGTCACATGGCATAATCAAA
Phenotype
MIM: 602187
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF195 PubMed Links

Gene Details

Gene
ZNF195
Gene Name
zinc finger protein 195
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130519.2 Intron NP_001123991.1
NM_001130520.2 Intron NP_001123992.1
NM_001242841.1 Intron NP_001229770.1
NM_001242842.1 Intron NP_001229771.1
NM_001242843.1 Intron NP_001229772.1
NM_001256823.1 Intron NP_001243752.1
NM_001256824.1 Intron NP_001243753.1
NM_001256825.1 Intron NP_001243754.1
NM_007152.4 Intron NP_009083.2
XM_006718306.1 Intron XP_006718369.1
XM_011520350.2 Intron XP_011518652.1
XM_011520351.2 Intron XP_011518653.1
XM_011520352.2 Intron XP_011518654.1
XM_011520354.2 Intron XP_011518656.2
XM_017018260.1 Intron XP_016873749.1
XM_017018261.1 Intron XP_016873750.1
XM_017018262.1 Intron XP_016873751.1
XM_017018263.1 Intron XP_016873752.1
XM_017018264.1 Intron XP_016873753.1
XM_017018265.1 Intron XP_016873754.1
XM_017018266.1 Intron XP_016873755.1
XM_017018267.1 Intron XP_016873756.1

View Full Product Details