Product Details

SNP ID
rs75224835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45806113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGCCCTGGTGCCGTGGACTT[A/C]CCCAGCTTGCGCCTGGACCTCAGGG
Phenotype
MIM: 605881
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC35C1 PubMed Links

Gene Details

Gene
SLC35C1
Gene Name
solute carrier family 35 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145265.1 1024 Silent Mutation TTA,TTC L91F NP_001138737.1
NM_001145266.1 1024 Silent Mutation TTA,TTC L91F NP_001138738.1
NM_018389.4 1024 Silent Mutation TTA,TTC L104F NP_060859.4
XM_011520202.2 1024 Intron XP_011518504.1
XM_011520203.2 1024 Missense Mutation TTA,TTC L104F XP_011518505.1

View Full Product Details