Product Details

SNP ID
rs76337493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6448881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATAAAGTGCAACCGTGGGGGT[A/G]GGGGTAGGAGAGGGAGGGCACCGGG
Phenotype
MIM: 142290 MIM: 605493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPX PubMed Links

Gene Details

Gene
HPX
Gene Name
hemopexin
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 2558 UTR 3 NP_001234935.1
NM_001248007.1 2558 UTR 3 NP_001234936.1
NM_006458.3 2558 UTR 3 NP_006449.2
NM_033278.3 2558 UTR 3 NP_150594.2
XM_011519844.1 2558 UTR 3 XP_011518146.1
XM_011519845.2 2558 UTR 3 XP_011518147.1
XM_017017098.1 2558 UTR 3 XP_016872587.1
XM_017017099.1 2558 UTR 3 XP_016872588.1

View Full Product Details