Product Details

SNP ID
rs78591962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122762234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTCTTTAATTTTTTTTTTTACC[A/T]TATGTATTTGAAGTTATTCTTGGTT
Phenotype
MIM: 604550
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DENR PubMed Links

Gene Details

Gene
DENR
Gene Name
density regulated re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003677.4 Intron NP_003668.2

View Full Product Details