Product Details

SNP ID
rs78179294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6557631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTACATTTGTAGGTGTGGCTG[G/T]TTCAGAATTTCCTGGGGTTAAAATT
Phenotype
MIM: 610495 MIM: 164031
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFFO1 PubMed Links

Gene Details

Gene
IFFO1
Gene Name
intermediate filament family orphan 1
There are no transcripts associated with this gene.

Gene
NOP2
Gene Name
NOP2 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033714.2 1919 Missense Mutation ACA,CCA T597P NP_001028886.1
NM_001258308.1 1919 Missense Mutation ACA,CCA T601P NP_001245237.1
NM_001258309.1 1919 Missense Mutation ACA,CCA T634P NP_001245238.1
NM_001258310.1 1919 UTR 3 NP_001245239.1
NM_006170.3 1919 Missense Mutation ACA,CCA T597P NP_006161.2
XM_005253691.1 1919 Missense Mutation ACA,CCA T601P XP_005253748.1

View Full Product Details