Product Details

SNP ID
rs79084421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121414270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATCTCGACTTCCATTACCTATC[A/C]CATTATCTAGATTCTAGAATATGCT
Phenotype
MIM: 609078 MIM: 608299
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KDM2B PubMed Links

Gene Details

Gene
KDM2B
Gene Name
lysine demethylase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005366.1 Intron NP_001005366.1
NM_032590.4 Intron NP_115979.3
XM_005253955.3 Intron XP_005254012.1
XM_005253956.3 Intron XP_005254013.1
XM_005253961.4 Intron XP_005254018.1
XM_011538867.2 Intron XP_011537169.1
XM_011538868.2 Intron XP_011537170.1
XM_011538869.2 Intron XP_011537171.1
XM_011538875.2 Intron XP_011537177.1
Gene
RNF34
Gene Name
ring finger protein 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256858.1 Intron NP_001243787.1
NM_025126.3 Intron NP_079402.2
NM_194271.2 Intron NP_919247.1

View Full Product Details