Product Details

SNP ID
rs77123766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111692760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCGTCTCCAGTGGGTGTGGAG[A/G]ACAGCCTTCCTGAAACACACCCAGC
Phenotype
MIM: 611181 MIM: 604986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAD10 PubMed Links

Gene Details

Gene
ACAD10
Gene Name
acyl-CoA dehydrogenase family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136538.1 251 Silent Mutation AGA,AGG R17R NP_001130010.1
NM_025247.5 251 Silent Mutation AGA,AGG R17R NP_079523.3
Gene
BRAP
Gene Name
BRCA1 associated protein
There are no transcripts associated with this gene.

View Full Product Details