Product Details

SNP ID
rs75218142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51755895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAAGAAAGGAGGGCACATGAA[C/T]CAAAGGACAAAAAGCAGGAAACCCG
Phenotype
MIM: 602928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD2 PubMed Links

Gene Details

Gene
LYSMD2
Gene Name
LysM domain containing 2
There are no transcripts associated with this gene.

Gene
TMOD2
Gene Name
tropomodulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142885.1 Intron NP_001136357.1
NM_014548.3 Intron NP_055363.1
XM_017022089.1 Intron XP_016877578.1

View Full Product Details