Product Details

SNP ID
rs80292319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76216291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTAGAGGCTAGGCATATTTC[C/T]GATAGTTTTAAAGCTGTGGAAAAGC
Phenotype
MIM: 608053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETFA PubMed Links

Gene Details

Gene
ETFA
Gene Name
electron transfer flavoprotein alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000126.3 1351 UTR 3 NP_000117.1
NM_001127716.1 1351 UTR 3 NP_001121188.1

View Full Product Details