Product Details

SNP ID
rs77368996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88878784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATGATTTATGCCCTGCATAACG[C/T]AGCAGCAGCAAAGCACTCAAAACTC
Phenotype
MIM: 155760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAN PubMed Links

Gene Details

Gene
ACAN
Gene Name
aggrecan
There are no transcripts associated with this gene.

Gene
HAPLN3
Gene Name
hyaluronan and proteoglycan link protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307952.1 Intron NP_001294881.1
NM_178232.3 Intron NP_839946.1
XM_011521261.1 Intron XP_011519563.1
XM_017021934.1 Intron XP_016877423.1
XM_017021935.1 Intron XP_016877424.1
XM_017021936.1 Intron XP_016877425.1

View Full Product Details