Product Details

SNP ID
rs75228366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:73093541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCACTCTCGGGCTGTCACACA[A/C]GCCTGGAGCCCAGCCATTTCTCGGC
Phenotype
MIM: 615613
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HCCAT5 PubMed Links

Gene Details

Gene
HCCAT5
Gene Name
hepatocellular carcinoma associated transcript 5 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details