Product Details

SNP ID
rs77806312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18042958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGTACTTTCTTTTCTGTGATT[C/T]TAGTGTAAAGAGTTCCTCAAACCTT
Phenotype
MIM: 608918
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATPAF2 PubMed Links

Gene Details

Gene
ATPAF2
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 2
There are no transcripts associated with this gene.

Gene
GID4
Gene Name
GID complex subunit 4 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024052.4 Intron NP_076957.3
XM_005256799.4 Intron XP_005256856.1
XM_005256800.1 Intron XP_005256857.1
XM_011524008.1 Intron XP_011522310.1

View Full Product Details