Product Details

SNP ID
rs79294240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18096873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTCCATTGCTCTGGGCCAGGCA[C/T]AGGGGATGGTGGTTAACAGCTACCT
Phenotype
MIM: 602986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRG2 PubMed Links

Gene Details

Gene
DRG2
Gene Name
developmentally regulated GTP binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001388.4 Intron NP_001379.1
XM_005256499.2 Intron XP_005256556.1
XM_005256500.2 Intron XP_005256557.1
XM_011523704.1 Intron XP_011522006.1
XM_011523705.1 Intron XP_011522007.1
XM_011523706.1 Intron XP_011522008.1
XM_011523707.1 Intron XP_011522009.1
XM_017024296.1 Intron XP_016879785.1
XM_017024297.1 Intron XP_016879786.1

View Full Product Details