Product Details

SNP ID
rs77340213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2323725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACAGCTGGTGTTGGAGTGGCTG[C/G]TGTGCTGTCTCAACATTTTCAAACT
Phenotype
MIM: 606477 MIM: 611214
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SNORD91A PubMed Links

Gene Details

Gene
SNORD91A
Gene Name
small nucleolar RNA, C/D box 91A
There are no transcripts associated with this gene.

Gene
SNORD91B
Gene Name
small nucleolar RNA, C/D box 91B
There are no transcripts associated with this gene.

Gene
SRR
Gene Name
serine racemase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304803.1 2164 Missense Mutation GCT,GGT A143G NP_001291732.1
NM_021947.2 2164 Missense Mutation GCT,GGT A292G NP_068766.1
XM_006721565.3 2164 Missense Mutation GCT,GGT A292G XP_006721628.1
XM_006721566.3 2164 Missense Mutation GCT,GGT A292G XP_006721629.1
XM_011523974.2 2164 Missense Mutation GCT,GGT A292G XP_011522276.1
Gene
TSR1
Gene Name
TSR1, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018128.4 2164 UTR 3 NP_060598.3

View Full Product Details