Product Details

SNP ID
rs75453384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9912511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAACGCCCAATACATAGCCAA[A/G]GACCGTGTGACACTGAAGCCTGGGT
Phenotype
MIM: 603127 MIM: 179618
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAS7 PubMed Links

Gene Details

Gene
GAS7
Gene Name
growth arrest specific 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130831.1 6220 UTR 3 NP_001124303.1
NM_003644.2 6220 UTR 3 NP_003635.2
NM_201432.1 6220 UTR 3 NP_958836.1
NM_201433.1 6220 UTR 3 NP_958839.1
XM_005256831.3 6220 UTR 3 XP_005256888.1
XM_005256832.3 6220 UTR 3 XP_005256889.1
XM_005256833.3 6220 UTR 3 XP_005256890.1
XM_005256834.3 6220 UTR 3 XP_005256891.1
XM_005256835.3 6220 UTR 3 XP_005256892.1
XM_011524044.2 6220 UTR 3 XP_011522346.1
XM_017025239.1 6220 UTR 3 XP_016880728.1
XM_017025240.1 6220 UTR 3 XP_016880729.1
XM_017025241.1 6220 UTR 3 XP_016880730.1
XM_017025242.1 6220 UTR 3 XP_016880731.1
Gene
RCVRN
Gene Name
recoverin
There are no transcripts associated with this gene.

View Full Product Details